MilliporeSigma
  • [4+2] cycloaddition reactions between 1,8-disubstituted cyclooctatetraenes and diazo dienophiles: stereoelectronic effects, anticancer properties and application to the synthesis of 7,8-substituted bicyclo[4.2.0]octa-2,4-dienes.
[4+2] cycloaddition reactions between 1,8-disubstituted cyclooctatetraenes and diazo dienophiles: stereoelectronic effects, anticancer properties and application to the synthesis of 7,8-substituted bicyclo[4.2.0]octa-2,4-dienes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-06-24)
Rebecca L Grange, Michael J Gallen, Heiko Schill, Jenny P Johns, Lin Dong, Peter G Parsons, Paul W Reddell, Victoria A Gordon, Paul V Bernhardt, Craig M Williams
ABSTRACT
A detailed examination of [4+2] cycloaddition reactions between 1,8-disubstituted cyclooctatetraenes and diazo compounds revealed that 4-phenyl-1,2,4-triazole-3,5-dione (PTAD) reacts to form either 2,3- or 3,4-disubstituted adducts. The product distribution can be controlled by modulating the electron density of the cyclooctatetraene. Unprecedented [4+2] cycloadditions between diisopropyl azodicarboxylate (DIAD) and 1,8-disubstituted cyclooctatetraenes are also described and further manipulation of a resulting cycloadduct uncovered a new pathway to the synthetically challenging bicyclo[4.2.0]octa-2,4-diene family. Variation of the substituents resulted in a range of compounds displaying selective action against different human tumour cell types.
Sigma-Aldrich
4-Phenyl-1,2,4-triazoline-3,5-dione, 97%