MilliporeSigma
  • Efficient asymmetric organocatalytic formation of erythrose and threose under aqueous conditions.
Efficient asymmetric organocatalytic formation of erythrose and threose under aqueous conditions.
Chemical communications (Cambridge, England) (2010-05-21)
Laurence Burroughs, Matthew E Vale, James A R Gilks, Henrietta Forintos, Christopher J Hayes, Paul A Clarke
ABSTRACT
Esters of proteinogenic amino acids efficiently catalyse the formation of erythrose and threose under aqueous conditions in the highest yields and enantioselectivities yet reported. Remarkably while esters of (L)-proline yield (L)-carbohydrates, esters of (L)-leucine and (L)-alanine generate (D)-carbohydrates, offering the potential to account for the prebiotic link between natural (L)-amino acids and natural (D)-sugars.