MilliporeSigma
  • A photosensing system composed of photosystem I, molecular wire, gold nanoparticle, and double surfactants in water.
A photosensing system composed of photosystem I, molecular wire, gold nanoparticle, and double surfactants in water.
Chemical communications (Cambridge, England) (2010-05-08)
Mariko Miyachi, Yoshinori Yamanoi, Yusuke Shibata, Hirokazu Matsumoto, Kazuo Nakazato, Masae Konno, Kohsuke Ito, Yasunori Inoue, Hiroshi Nishihara
ABSTRACT
Photosensing performance of a system composed of photosystem I (PSI), vitamin K(1) (VK(1))-like molecular wire, and gold nanoparticles (AuNPs) in an aqueous solution was increased considerably by the addition of double surfactants, hexylamine and dodecylbenzenesulfonate.
Sigma-Aldrich
Hexylamine, 99%