MilliporeSigma
  • N-Methyl stereochemistry in tropinone: the conformational flexibility of the tropane motif.
N-Methyl stereochemistry in tropinone: the conformational flexibility of the tropane motif.
Physical chemistry chemical physics : PCCP (2010-04-21)
Emilio J Cocinero, Alberto Lesarri, Patricia Ecija, Jens-Uwe Grabow, José A Fernández, Fernando Castaño
ABSTRACT
The intrinsic conformational and structural properties of the tropinone azabicycle have been investigated in a supersonic jet expansion using rotational spectroscopy. The spectrum revealed the presence of equatorial and axial conformers originated by the inversion of the N-methyl group, with the tropane motif adopting a distorted chair configuration. The determination of substitution and effective structures for the two conformers reveals the flexibility and structural changes associated with the N-methyl inversion, mostly a flattening at the nitrogen atom and a simultaneous rising of the carbonyl group in the axial form. Relative intensity measurements indicate that the conformational equilibrium is displaced towards the equatorial form, with a relative population in the jet of N(eq)/N(ax) approximately 2/1, which would correspond to a relative energy of ca. 2 kJ mol(-1) in pre-expansion conditions. Supporting ab initio calculations of the molecular properties and inversion barrier complemented the experimental work.
Sigma-Aldrich
Tropinone, 99%