MilliporeSigma
  • Effect of 1-aminoanthracene (1-AMA) binding on the structure of three lipocalin proteins, the dimeric β lactoglobulin, the dimeric odorant binding protein and the monomeric α1-acid glycoprotein. Fluorescence spectra and lifetimes studies.
Effect of 1-aminoanthracene (1-AMA) binding on the structure of three lipocalin proteins, the dimeric β lactoglobulin, the dimeric odorant binding protein and the monomeric α1-acid glycoprotein. Fluorescence spectra and lifetimes studies.
Journal of fluorescence (2010-03-31)
Daniel Kmiecik, Jihad René Albani
ABSTRACT
We studied effect of 1-aminoanthracene (1-AMA) binding on the structures of dimeric β lactoglobulin, dimeric odorant binding protein (OBP) and monomeric α(1)-acid glycoprotein (lipocalin family proteins) by monitoring fluorescence excitation spectra and measuring fluorescence lifetimes of the tryptophan residues of the proteins. Results show that binding of 1-AMA to β lactoglobulin and OBP modifies their conformation even at low probe concentration compared to that of the proteins. Structural modification induces a red shift of the fluorescence excitation spectra maximum of tryptophan residues accompanied with an increase of the third fluorescence lifetime and a decrease of its pre-exponential factor. These effects were not observed for α(1)-acid glycoprotein, probably as the result of carbohydrate presence. These data raise doubts concerning use of 1-AMA as a probe to study biological properties of β lactoglobulin and OBP.
Sigma-Aldrich
1-Aminoanthracene, technical grade, 90%