MilliporeSigma
  • [Absolute quantification of carminic acid in cochineal extract by quantitative NMR].
[Absolute quantification of carminic acid in cochineal extract by quantitative NMR].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2010-03-09)
Naoki Sugimoto, Atsuko Tada, Takako Suematsu, Kazunori Arifuku, Takeshi Saito, Toshihide Ihara, Yuuichi Yoshida, Reiji Kubota, Maiko Tahara, Kumiko Shimizu, Sumio Ito, Takeshi Yamazaki, Yoko Kawamura, Tetsuji Nishimura
ABSTRACT
A quantitative NMR (qNMR) method was applied for the determination of carminic acid. Carminic acid is the main component in cochineal dye that is widely used as a natural food colorant. Since several manufacturers only provide reagent-grade carminic acid, there is no reference material of established purity. To improve the reliability of analytical data, we are developing quantitative nuclear magnetic resonance (qNMR), based on the fact that the intensity of a given NMR resonance is directly proportional to the molar amount of that nucleus in the sample. The purities and contents of carminic acid were calculated from the ratio of the signal intensities of an aromatic proton on carminic acid to nine protons of three methyl groups on DSS-d6 used as the internal standard. The concentration of DSS-d6 itself was corrected using potassium hydrogen phthalate, which is a certified reference material (CRM). The purities of the reagents and the contents of carminic acid in cochineal dye products were determined with SI-traceability as 25.3-92.9% and 4.6-30.5% based on the crystalline formula, carminic acid potassium salt trihydrate, which has been confirmed by X-ray analysis. The qNMR method does not require a reference compound, and is rapid and simple, with an overall analysis time of only 10 min. Our approach thus represents an absolute quantitation method with SI-traceability that should be readily applicable to analysis and quality control of any natural product.
Product Description
Supelco
Carminic acid, analytical standard
Sigma-Aldrich
Carminic acid, ≥90% (HPLC)