MilliporeSigma
  • Improved methodology for identifying the teratogenic potential in early drug development of hERG channel blocking drugs.
Improved methodology for identifying the teratogenic potential in early drug development of hERG channel blocking drugs.
Reproductive toxicology (Elmsford, N.Y.) (2010-02-11)
M F Nilsson, C Danielsson, A-C Sköld, A Johansson, B Blomgren, J Wilson, K M Khan, E Bengtsson, K Kultima, W S Webster, B R Danielsson
ABSTRACT
Drugs blocking the potassium current IKr of the heart (via hERG channel-inhibition) have the potential to cause hypoxia-related teratogenic effects. However, this activity may be missed in conventional teratology studies because repeat dosing may cause resorptions. The aim of the present study was to investigate an alternative protocol to reveal the teratogenic potential of IKr-blocking drugs. The IKr blocker astemizole, given as a single dose (80 mg/kg) on gestation day (GD) 13 to pregnant rats caused digital defects. In whole rat embryo culture (2h) on GD 13, astemizole caused a decrease in embryonic heart rate at 20 nM, and arrhythmias at 200-400 nM. Cetirizine, without IKr-blocking properties, did not affect the rat embryonic heart in vitro. The present study shows that single dose testing on sensitive days of development, together with whole embryo culture, can be a useful methodology to better characterize the teratogenic potential of IKr-blocking drugs.
Sigma-Aldrich
Astemizole, ≥98% (HPLC)
Pricing and availability is not currently available.