MilliporeSigma
  • Involvement of Ca2+ and ROS in alpha-tocopheryl succinate-induced mitochondrial permeabilization.
Involvement of Ca2+ and ROS in alpha-tocopheryl succinate-induced mitochondrial permeabilization.
International journal of cancer (2010-01-28)
Vladimir Gogvadze, Erik Norberg, Sten Orrenius, Boris Zhivotovsky
ABSTRACT
Release of mitochondrial proteins such as cytochrome c, AIF, Smac/Diablo etc., plays a crucial role in apoptosis induction. A redox-silent analog of vitamin E, alpha-tocopheryl succinate (alpha-TOS), was shown to stimulate cytochrome c release via production of reactive oxygen species (ROS) and Bax-mediated permeabilization of the outer mitochondrial membrane. Here we show that alpha-TOS facilitates mitochondrial permeability transition (MPT) in isolated rat liver mitochondria, Tet21N neuroblastoma cells and Jurkat T-lymphocytes. In particular, in addition to ROS production, alpha-TOS stimulates rapid Ca(2+) entry into the cells with subsequent accumulation of Ca(2+) in mitochondria-a prerequisite step for MPT induction. Alteration of mitochondrial Ca(2+) buffering capacity was observed as early as 8 hr after incubation with alpha-TOS, when no activation of Bax was yet detected. Ca(2+) accumulation in mitochondria was important for apoptosis progression, since inhibition of mitochondrial Ca(2+) uptake significantly mitigated the apoptotic response. Importantly, Ca(2+)-induced mitochondrial destabilization might cooperate with Bax-mediated mitochondrial outer membrane permeabilization to induce cytochrome c release from mitochondria.
Sigma-Aldrich
D-α-Tocotrienol, from rice, ≥75% (GC)