MilliporeSigma
Novel synthesis of ureas: application of t-butylureas.
Chemical & pharmaceutical bulletin (2010-01-05)
Ai Ito, Hideaki Muratake, Koichi Shudo
ABSTRACT
Otherwise inaccessible tropolonylureas were prepared by reaction of t-butylurea with appropriate amines, with elimination of t-butylamine. This method is also generally applicable for urea synthesis.
Sigma-Aldrich
N-Butylurea, 99%