MilliporeSigma
  • Extended O6-methylguanine methyltransferase promoter hypermethylation following n-butylidenephthalide combined with 1,3-bis(2-chloroethyl)-1-nitrosourea (BCNU) on inhibition of human hepatocellular carcinoma cell growth.
Extended O6-methylguanine methyltransferase promoter hypermethylation following n-butylidenephthalide combined with 1,3-bis(2-chloroethyl)-1-nitrosourea (BCNU) on inhibition of human hepatocellular carcinoma cell growth.
Journal of agricultural and food chemistry (2010-01-02)
Yung-Luen Yu, Sung-Liang Yu, Kuo-Jung Su, Chyou-Wei Wei, Min-Hui Jian, Po-Cheng Lin, I-Hsuan Tseng, Chai-Ching Lin, Chin-Cheng Su, De-Chuan Chan, Shinn-Zong Lin, Horng-Jyh Harn, Yi-Lin Sophia Chen
ABSTRACT
Epigenetic alteration of DNA methylation plays an important role in the regulation of gene expression associated with chemosensitivity of human hepatocellular (HCC) carcinoma cells. With the aim of improving the chemotherapeutic efficacy for HCC, the effect of the naturally occurring compound n-butylidenephthalide (BP), which is isolated from a chloroform extract of Angelica sinensis, was investigated. In both HepG2 and J5 HCC cell lines, a synergistic antiproliferative effect was observed when a low dosage of BP was combined with the chemotherapeutic drug 1,3-bis(2-chloroethyl)-1-nitrosourea (BCNU). BCNU is an alkylating agent, and it prompts us to examine one of DNA repair genes, O(6)-methylguanine methyltransferase (MGMT). It was evident from methylation-specific polymerase chain reaction (PCR) analysis that BP/BCNU combined treatment caused a time- and concentration-dependent enhancement of MGMT promoter methylation. Overexpression of MGMT could abolish BP-induced growth inhibition in the J5 tumor cell line as measured by colony formation assay. When BP was combined with BCNU and administered, it showed significant antitumor effects in both HepG2 and J5 xenograft tumors as compared with the use of only one of these drugs. The BCNU-induced apoptosis and inhibited MGMT protein expression in HCC cells, both in vitro and in vivo, resulting from the combination treatment of BP and BCNU suggest a potential clinical use of this compound for improving the prognosis for HCCs.