MilliporeSigma
  • Enzymatic production of (-)-indolactam V by LtxB, a cytochrome P450 monooxygenase.
Enzymatic production of (-)-indolactam V by LtxB, a cytochrome P450 monooxygenase.
Journal of natural products (2009-12-17)
Minh U Huynh, Matthew C Elston, Nick M Hernandez, David B Ball, Shin-ichiro Kajiyama, Kazuhiro Irie, William H Gerwick, Daniel J Edwards
ABSTRACT
The P450 cytochrome monooxygenase gene, ltxB, was cloned and overexpressed in Escherichia coli as a 6xHis-tagged protein. The resulting recombinant LtxB was purified by Ni-NTA affinity chromatography and characterized biochemically. Purified LtxB demonstrated typical cytochrome P450 spectroscopic properties including substrate-induced transition from a low-spin (lambdamax=414 nm) to high-spin state (lambdamax=386 nm) upon incubation with N-methyl-L-valyl-L-tryptophanol. The catalytic activity of LtxB was verified by demonstrating the oxidation/cyclization of N-methyl-L-valyl-L-tryptophanol to (-)-indolactam V. LtxB shows a relaxed specificity for analogue substrates in which the valyl group is substituted for other aliphatic groups. The relaxed substrate specificity of LtxB, along with the relaxed specificity of the prenyltransferase, LtxC, allowed for the enzymatic production of a series of (-)-indolactam V and lyngbyatoxin analogues.
Sigma-Aldrich
(−)-Indolactam V, ≥96% (HPLC)