MilliporeSigma
Plant-based corosolic acid: future anti-diabetic drug?
Biotechnology journal (2009-12-01)
Ganapathy Sivakumar, Daniel R Vail, Vipin Nair, Fabricio Medina-Bolivar, Jackson O Lay
ABSTRACT
Diabetes is one of the nation's most prevalent, debilitating and costly diseases. For diabetes, frequent insulin treatment is very expensive and may increase anti-insulin antibody production, which may cause unwanted side effects. Corosolic acid may also have some efficacy in the treatment of diabetes, but without induction of anti-insulin antibodies. Recently, corosolic acid from Lagerstroemia speciosa L. leaf extracts has been reported to act via an indirect mechanism (unlike insulin) in animal experiments. The insulin-complementary anti-diabetic therapeutic value observed in these Japanese preliminary clinical trials has led to renewed interest in the biosynthesis of this compound. So far, there has been no clear evidence for a corosolic acid biosynthetic pathway in plants. This article provides possible roles of corosolic acid and hypothetical information on the biosynthetic pathway in plants.
Sigma-Aldrich
Corosolic acid from Lagerstroemia speciosa, ≥98% (HPLC)
Supelco
Corosolic acid, analytical standard