MilliporeSigma
  • In vitro stereoselective hydrolysis of diacylglycerols by hormone-sensitive lipase.
In vitro stereoselective hydrolysis of diacylglycerols by hormone-sensitive lipase.
Biochimica et biophysica acta (2009-10-06)
Jorge A Rodriguez, Yassine Ben Ali, Slim Abdelkafi, Lilia D Mendoza, Julien Leclaire, Frédéric Fotiadu, Gerard Buono, Frédéric Carrière, Abdelkarim Abousalham
ABSTRACT
Hormone-sensitive lipase (HSL) contributes importantly to the mobilization of fatty acids in adipocytes and shows a substrate preference for the diacylglycerols (DAGs) originating from triacylglycerols. To determine whether HSL shows any stereopreference during the hydrolysis of diacylglycerols, racemic 1,2(2,3)-sn-diolein was used as a substrate and the enantiomeric excess (ee%) of residual 1,2-sn-diolein over 2,3-sn-diolein was measured as a function of DAG hydrolysis. Enantiomeric DAGs were separated by performing chiral-stationary-phase HPLC after direct derivatization from lipolysis product extracts. The fact that the ee% of 1,2-sn-diolein over 2,3-sn-diolein increased with the level of hydrolysis indicated that HSL has a preference for 2,3-sn-diolein as a substrate and therefore a stereopreference for the sn-3 position of dioleoylglycerol. The ee% of 1,2-sn-diolein reached a maximum value of 36% at 42% hydrolysis. Among the various mammalian lipases tested so far, HSL is the only lipolytic carboxylester hydrolase found to have a pronounced stereospecificity for the sn-3 position of dioleoylglycerol.
Sigma-Aldrich
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid