MilliporeSigma
Effects of tellurite on growth of Saccharomyces cerevisiae.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2009-09-18)
Domenica R Massardo, Paola Pontieri, Loredana Maddaluno, Mario De Stefano, Pietro Alifano, Luigi Del Giudice
ABSTRACT
The effects of potassium tellurite on growth and survival of rho(+) and rho(0) Saccharomyces cerevisiae strains were investigated. Both rho(+) and rho(0) strains grew on a fermentable carbon source with up to 1.2 mM K(2)TeO(3), while rho(+) yeast cells grown on a non-fermentable carbon source were inhibited at tellurite levels as low as 50 muM suggesting that this metalloid specifically inhibited mitochondrial functions. Growth of rho(+) yeast cells in the presence of increasing amount of tellurite resulted in dose-dependent blackening of the culture, a phenomenon not observed with rho(0) cultures. Transmission electron microscopy of S. cerevisiae rho(+) cells grown in the presence of tellurite showed that blackening was likely due to elemental tellurium (Te(0)) that formed large deposits along the cell wall and small precipitates in both the cytoplasm and mitochondria.
Product Description
Millipore
Potassium tellurite solution, 1% in H2O, suitable for microbiology