MilliporeSigma
  • A suitable preparation of N-sulfonyl-1,2,3,4-tetrahydroisoquinolines and their ring homologs with a reusable Preyssler heteropolyacid as catalyst.
A suitable preparation of N-sulfonyl-1,2,3,4-tetrahydroisoquinolines and their ring homologs with a reusable Preyssler heteropolyacid as catalyst.
Molecular diversity (2009-07-03)
Gustavo P Romanelli, Diego M Ruiz, Juan C Autino, Héctor E Giaccio
ABSTRACT
The preparation of N-sulfonyl-1,2,3,4-tetrahydroisoquinolines, N-sulfonyl-2,3,4,5-tetrahydro-1H-2-benzazepines and N-sulfonyl-1,2,3,4,5,6-hexahydrobenzazocine was catalyzed by a Preyssler heteropolyacid, H(14)[NaP(5)W(30)O(110)], (PA), supported on silica (PASiO(2)40) with excellent yields by means of the Pictet-Spengler reaction of N-aralkylsulfonamides with s-trioxane. The reactions proceed with 0.5 mol% of silica-supported catalyst in toluene at 70°C. The catalyst can be recycled without appreciable loss of the catalytic activity.
Sigma-Aldrich
1,2,3,4-Tetrahydroisoquinoline, 95%