MilliporeSigma
Acid-base actuation of [c2]daisy chains.
Journal of the American Chemical Society (2009-05-08)
Lei Fang, Mohamad Hmadeh, Jishan Wu, Mark A Olson, Jason M Spruell, Ali Trabolsi, Ying-Wei Yang, Mourad Elhabiri, Anne-Marie Albrecht-Gary, J Fraser Stoddart
ABSTRACT
A versatile synthetic strategy, which was conceived and employed to prepare doubly threaded, bistable [c2]daisy chain compounds, is described. Propargyl and 1-pentenyl groups have been grafted onto the stoppers of [c2]daisy chain molecules obtained using a template-directed synthetic protocol. Such [c2]daisy chain molecules undergo reversible extension and contraction upon treatment with acid and base, respectively. The dialkyne-functionalized [c2]daisy chain (AA) was subjected to an [AA+BB] type polymerization with an appropriate diazide (BB) to afford a linear, mechanically interlocked, main-chain polymer. The macromolecular properties of this polymer were characterized by chronocoulometry, size exclusion chromatography, and static light-scattering analysis. The acid-base switching properties of both the monomers and the polymer have been studied in solution, using (1)H NMR spectroscopy, UV/vis absorption spectroscopy, and cyclic voltammetry. The experimental results demonstrate that the functionalized [c2]daisy chains, along with their polymeric derivatives, undergo quantitative, efficient, and fully reversible switching processes in solution. Kinetics measurements demonstrate that the acid/base-promoted extension/contraction movements of the polymeric [c2]daisy chain are actually faster than those of its monomeric counterpart. These observations open the door to correlated molecular motions and to changes in material properties.
Sigma-Aldrich
Dibenzo-24-crown-8, 98%