MilliporeSigma
  • Efficacy of salvage radiotherapy plus 2-year androgen suppression for postradical prostatectomy patients with PSA relapse.
Efficacy of salvage radiotherapy plus 2-year androgen suppression for postradical prostatectomy patients with PSA relapse.
International journal of radiation oncology, biology, physics (2009-05-05)
Richard Choo, Cyril Danjoux, Sandra Gardner, Gerard Morton, Ewa Szumacher, D Andrew Loblaw, Patrick Cheung, Maria Pearse
ABSTRACT
To determine the efficacy of a combined approach of radiotherapy (RT) plus 2-year androgen suppression (AS) as salvage treatment for prostate-specific antigen (PSA) relapse after radical prostatectomy (RP). Seventy-five patients with PSA relapse after RP were treated with salvage RT plus 2-year AS, as per a pilot, prospective study. AS started within 1 month after completion of salvage RT and consisted of nilutamide for 4 weeks and buserelin acetate depot subcutaneously every 2 months for 2 years. Relapse-free rate including freedom from PSA relapse was estimated using the Kaplan-Meier method. PSA relapse was defined as a PSA rise above 0.2 ng/mL with two consecutive increases over a minimum of 3 months. A Cox regression analysis was performed to evaluate prognostic factors for relapse. Median age of the cohort was 63 years at the time of salvage RT. Median follow-up from salvage RT was 6.4 years. All achieved initially complete PSA response (< 0.2) with the protocol treatment. Relapse-free rate including the freedom from PSA relapse was 91.5% at 5 years and 78.6% at 7 years. Overall survival rate was 93.2% at both 5 and 7 years. On Cox regression analysis, pT3 stage and PSA relapse less than 2 years after RP were significant prognostic factors for relapse. The combined treatment of salvage RT plus 2-year AS yielded an encouraging result for patients with PSA relapse after RP and needs a confirmatory study.
Product Description
Nilutamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nilutamide, solid