MilliporeSigma
  • Enhancement of potent antibody and T-cell responses by a single-dose, novel nanoemulsion-formulated pandemic influenza vaccine.
Enhancement of potent antibody and T-cell responses by a single-dose, novel nanoemulsion-formulated pandemic influenza vaccine.
Microbes and infection (2009-04-07)
Ming-Hsi Huang, Chiung-Yi Huang, Su-Chen Lin, Jia-Huey Chen, Chien-Chun Ku, Ai-Hsiang Chou, Shih-Jen Liu, Hsin-Wei Chen, Pele Chong, Chih-Hsiang Leng
ABSTRACT
Vaccine shortages are a major obstacle to influenza pandemic preparedness. Increasing vaccine efficiency provides a potentially effective way to overcome this problem. Specifically, using single-dose immunization to induce protective immunity is an attractive approach to emergency/massive vaccination. In this report, we propose a novel nanoemulsion comprised of the bioresorbable polymer, Span 85, and squalene forming a ready-to-use adjuvant, called PELC. After formulation with PELC, inactivated H5N1 virus was intramuscularly administered to mice via a single injection. The data demonstrate that inactivated virus containing 0.5microg hemagglutinin (HA) and formulated with PELC induced more potent antigen-specific antibodies, hemagglutination inhibition, and virus neutralization than non-adjuvanted inactivated virus containing 5microg HA. In addition, T-cell proliferative responses, as well as interferon-gamma (IFN-gamma) and interleukin-4 (IL-4) secretion were significantly enhanced after immunization with PELC-adjuvanted inactivated virus. These results indicate that PELC can be used for effective single-dose immunization and could thus play an important role in influenza pandemic preparedness.
Sigma-Aldrich
Span® 85
Pricing and availability is not currently available.