MilliporeSigma
  • Detection thresholds for 2-isopropyl-3-methoxypyrazine in Concord and Niagara grape juice.
Detection thresholds for 2-isopropyl-3-methoxypyrazine in Concord and Niagara grape juice.
Journal of food science (2009-02-26)
G J Pickering, A Karthik, D Inglis, M Sears, K Ker
ABSTRACT
2-isopropyl-3-methoxypyrazine (IPMP) is the compound responsible for the off-flavor known as ladybug taint, which occurs when Harmnonia axyridis beetles become incorporated with the grapes during juice processing. It is also an important grape-derived component of juice flavor in some varieties. The main objective of this study was to determine the orthonasal (ON) and retronasal (RN) detection thresholds for IPMP in juice. The ASTM E679 ascending forced choice method of limits was used to determine detection thresholds for 26 individuals in Concord and Niagara juices. Group best estimate thresholds (BETs) averaged 0.93 ng/L and were 50% and 21% higher in Concord than in Niagara juices for ON and RN evaluation, respectively. Group BETs for IPMP (ng/L) for Concord were ON: 1.11; RN: 1.02 and for Niagara were ON: 0.74; RN: 0.84. Variation in individual detection thresholds was observed, although familiarity with ladybug taint was not associated with individual threshold values. We conclude that humans are very sensitive to IPMP in juice, and that detection thresholds are more strongly influenced by grape variety than evaluation mode. These results may assist juice producers in establishing tolerance levels for IPMP in juice affected by ladybug taint or derived from grapes of suboptimal ripeness.
Product Description
Supelco
2-Isopropyl-3-methoxypyrazine solution, certified reference material
Pricing and availability is not currently available.
Sigma-Aldrich
2-Isopropyl-3-methoxypyrazine, 97%
Pricing and availability is not currently available.