MilliporeSigma
Photovalorisation of pentafluorobenzoic acid with platinum doped TiO2.
Journal of hazardous materials (2009-02-13)
L Ravichandran, K Selvam, B Krishnakumar, M Swaminathan
ABSTRACT
Pt-doped TiO(2) has been prepared and characterized by various surface analytical methods such as BET surface area, scanning electron micrographs (SEM), X-ray diffraction (XRD), energy dispersive X-ray micro analysis (EDX) and diffuse reflectance spectroscopy (DRS). Photodefluoridation of pentafluorobenzoic acid (PFBA) to fluoride ions has been carried out using TiO(2) and Platinum doped TiO(2) with UV-C (200-300 nm) light. The defluoridation was monitored by the ionometer with fluoride ion selective electrode. Photodeposition of 1.5% Platinum on the surface of TiO(2) enhances its photoactivity in pentafluorobenzoic acid defluoridation. In order to optimize the working conditions, effects of pH and initial PFBA concentration were analyzed. Addition of inorganic oxidizing species KIO(4,) H(2)O(2), (NH(4))S(2)O(8) and KBrO(3) strongly influenced the photocatalytic defluoridation of PFBA. The significant enhancement in the Pt-TiO(2) photoactivity under UV irradiation can be ascribed to the platinum deposits, acting as electron traps on the Pt-TiO(2) surface. The fluoride ions formed during photodefluoridation can be used for the production of CaF(2).
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzoic acid, 99%