MilliporeSigma
  • In vitro evaluation of dentifrice with low fluoride content supplemented with trimetaphosphate.
In vitro evaluation of dentifrice with low fluoride content supplemented with trimetaphosphate.
Caries research (2009-01-29)
E M Takeshita, L P Castro, K T Sassaki, A C B Delbem
ABSTRACT
One way to reduce dental fluorosis is by reducing the fluoride (F) concentration in dentifrice, but low-F dentifrice should be as effective as a standard dentifrice. This study evaluated in vitro whether the supplementation with sodium trimetaphosphate (TMP) of a dentifrice with low F content (500 microg/g) would provide a similar effect to that of a standard dentifrice. Bovine enamel blocks were submitted to a pH cycling regime incorporating daily exposures to a slurry of dentifrice: a low-F dentifrice with or without 0.1-3.0% TMP; an F-free, phosphate-free dentifrice (negative control), or a dentifrice with 1,100 microg/g F (positive control). The addition of TMP to dentifrice with or without F was associated with higher surface hardness and decreased loss of integrated subsurface hardness after pH cycling. The combination of 1% TMP and 500 microg F/g had a greater effect than the positive control dentifrice. It is concluded that the addition of TMP to the 500-microg F/g dentifrice allowed a similar or larger effect as compared with a standard dentifrice in this in vitro model.
Sigma-Aldrich
Trisodium trimetaphosphate, ≥95%