MilliporeSigma
  • In vitro study on the interaction between thiophanate methyl and human serum albumin.
In vitro study on the interaction between thiophanate methyl and human serum albumin.
Journal of photochemistry and photobiology. B, Biology (2009-01-06)
Jinhua Li, Xiaoyan Liu, Cuiling Ren, Jiazhong Li, Fenling Sheng, Zhide Hu
ABSTRACT
Thiophanate methyl (MT) is one of the widely used fungicides to control important fungal diseases of crops, which has led to potential toxicological risk to public health. Several different transport proteins exist in blood plasma, but albumin only is bound by a wide diversity of xenobiotics reversibly with high affinity. We studied the interaction of MT with human serum albumin by using spectroscopic methods including fluorescence quenching technology, UV and Fourier transform infrared (FT-IR) spectroscopy under simulative physiological conditions. The result of fluorescence titration revealed that MT could quench the intrinsic fluorescence of HSA. The binding process was exothermic and spontaneous, as indicated by the thermodynamic analyses. In addition, the studies of FT-IR spectroscopy showed that the binding of MT to HSA changed molecular conformation of HSA. The results obtained from molecular modeling showed that the interaction between MT and HSA was dominated by hydrophobic force, and there was also hydrogen bond interaction between the pesticide and the residues of HSA, which was in good agreement with the result of binding mode.
Product Description
Supelco
Thiophanate-methyl, PESTANAL®, analytical standard