MilliporeSigma
  • Hexachlorophene suppresses beta-catenin expression by up-regulation of Siah-1 in EBV-infected B lymphoma cells.
Hexachlorophene suppresses beta-catenin expression by up-regulation of Siah-1 in EBV-infected B lymphoma cells.
Cancer letters (2008-12-19)
Hye-Jin Min, Il-Rae Cho, Ratakorn Srisuttee, Eun-Hee Park, Dae Ho Cho, Jin-Hyun Ahn, Im-Soon Lee, Randal N Johnston, Sangtaek Oh, Young-Hwa Chung
ABSTRACT
Many studies have shown that the activation of beta-catenin signaling can promote oncogenesis, and it is therefore of interest to find agents that modulate this pathway. Recent work has shown using B lymphoma cells that infection by Epstein-Barr virus (EBV) and expression of its latent membrane protein (LMP)-1, cause increases in the expression of beta-catenin and cellular transformation. Conversely, results from cell-based small molecule screening studies have shown that the antibiotic hexachlorophene can down-regulate beta-catenin in colon cancer cells. Here we report that hexachlorophene also counteracts the elevated beta-catenin levels in EBV-infected B lymphomas. This is associated with restoration in levels of Siah-1 (an E3 ubiquitin ligase that is active in beta-catenin regulation) which had been diminished by LMP-1. Our results suggest that Siah-1 is targeted by both LMP-1 and hexachlorophene with opposite effects. The hexachlorophene modulation of Siah-1 and beta-catenin is independent of p53 and results in reduced expression of cyclin-D1 and c-Myc (target genes of beta-catenin), leading to the growth arrest of B lymphoma cells. From these results we propose that hexachlorophene may provide a novel therapeutic strategy for EBV-infected B lymphoma cells by reducing beta-catenin levels via the restoration of Siah-1.
Product Description
Supelco
Hexachlorophene, PESTANAL®, analytical standard