MilliporeSigma
  • A one-pot oxidative decarboxylation-Friedel-Crafts reaction of acyclic alpha-amino acid derivatives activated by the combination of iodobenzene diacetate/iodine and iron dust.
A one-pot oxidative decarboxylation-Friedel-Crafts reaction of acyclic alpha-amino acid derivatives activated by the combination of iodobenzene diacetate/iodine and iron dust.
Organic & biomolecular chemistry (2008-11-29)
Renhua Fan, Weixun Li, Bing Wang
ABSTRACT
An efficient one-pot oxidative decarboxylation-Friedel-Crafts reaction of acyclic alpha-amino acid derivatives with electron-rich aromatic compounds is reported. The reaction is activated by the combination of iodobenzene diacetate, iodine and iron dust, resulting in a mild and simple reaction system. The use of iron avoids the oxidation of aromatic compounds, and in situ generation of Fe(III) salts to promote the Friedel-Crafts reaction avoids the use of the highly hygroscopic FeCl(3).
Sigma-Aldrich
(Diacetoxyiodo)benzene, 98%
Pricing and availability is not currently available.