MilliporeSigma
L-ornithine-L-aspartate infusion efficacy in hepatic encephalopathy.
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP (2008-11-06)
Irfan Ahmad, Anwaar A Khan, Altaf Alam, Akif Dilshad, Arshad Kamal Butt, Farzana Shafqat, Kashif Malik, Shahid Sarwar
ABSTRACT
To determine the efficacy of L-ornithine-L-aspartate in treatment of hepatic encephalopathy. Randomized, placebo-controlled trial. Department of Gastroenterology and Hepatology, Sheikh Zayed Hospital, Lahore, from February to August 2005. Cirrhotic patients with hyperammonemia and overt hepatic encephalopathy were enrolled. Eighty patients were randomized to two treatment groups, L-ornithine-L-aspartate (20 g/d) or placebo, both dissolved in 250 mL of 5% dextrose water and infused intravenously for four hours a day for five consecutive days with 0.5 g/kg dietary protein intake at the end of daily treatment period. Outcome variables were postprandial blood ammonia and mental state grade. Adverse reactions and mortality were also determined. Both treatment groups were comparable regarding age, gender, etiology of cirrhosis, Child-Pugh class, mental state grade and blood ammonia at baseline. Although, improvement occurred in both groups, there was a greater improvement in L-ornithine-L-aspartate group with regard to both variables. Four patients in the placebo group and 2 in L-ornithine-L-aspartate group died. L-ornithine-L-aspartate infusions were found to be effective in cirrhotic patients with hepatic encephalopathy.
Sigma-Aldrich
L-Ornithine L-aspartate salt, powder