MilliporeSigma
  • Hydrogen-bond patterns and the structures of 1,4-cyclohexanediol: 2:1 cis:trans-1,4-cyclohexanediol.
Hydrogen-bond patterns and the structures of 1,4-cyclohexanediol: 2:1 cis:trans-1,4-cyclohexanediol.
Acta crystallographica. Section B, Structural science (2008-09-19)
James H Loehlin, Michelle Lee, Christina M Woo
ABSTRACT
The structure of a co-crystal with both trans and cis isomers of 1,4-cyclohexanediol (1,4-CHD) is reported. The intermolecular hydrogen-bond patterns are described and compared with those of the all trans structure, using the graph-set model. A second crystal with possible cis/trans disorder is also described. The results of molecular modeling of the simple isomers and conformers are compared with the known structures.
Sigma-Aldrich
1,4-Cyclohexanediol, 99%
Pricing and availability is not currently available.