MilliporeSigma
  • Enzyme assay and activity fingerprinting of hydrolases with the red-chromogenic adrenaline test.
Enzyme assay and activity fingerprinting of hydrolases with the red-chromogenic adrenaline test.
Nature protocols (2008-08-21)
Viviana S Fluxá, Denis Wahler, Jean-Louis Reymond
ABSTRACT
The adrenaline test for enzymes is a colorimetric enzyme assay based on the quantification of periodate-sensitive reaction products such as 1,2-diols and 1,2-aminoalcohols by back-titration of the oxidant with adrenaline to produce adrenochrome as an easily detectable red product. The test uses commercial reagents and is suitable for screening the activity of various hydrolases. It is demonstrated here for testing epoxide hydrolases, lipases and esterases, and for activity fingerprinting of these enzymes across substrate series. The complete assay requires 2-3 h.
Sigma-Aldrich
Adrenochrome, ≥95% (HPLC)