MilliporeSigma
Effect of scoparone on neurite outgrowth in PC12 cells.
Neuroscience letters (2008-06-13)
Yoo Jung Yang, Hak Ju Lee, Don Ha Choi, Hai Shan Huang, Sung Cil Lim, Myung Koo Lee
ABSTRACT
The neurite outgrowth-promoting effects of scoparone isolated from the stem bark of Liriodendron tulipifera were investigated in PC12 cells. At a concentration of 200 microM, scoparone markedly induced neurite outgrowth from PC12 cells. Scoparone at 200 microM also enhanced the outgrowth of neurites from cells in the presence of nerve growth factor (NGF, 2 ng/ml). The levels of intracellular cyclic AMP and concentration of Ca2+ were also increased by 200 microM scoparone. In addition, scoparone at 200 microM increased the activities of extracellular signal-regulated protein kinase (ERK), cyclic AMP-dependent protein kinase (PKA), protein kinase C (PKC) and Ca2+/calmodulin kinase II (CaMK II). However, scoparone-induced neurite outgrowth was blocked by a mitogen-activated protein kinase inhibitor (U0126), a PKA inhibitor (H89), a PKC inhibitor (GF109203X) and a CaMK II inhibitor (KN62). These kinase inhibitors also reduced the scoparone-induced neurite outgrowth associated with NGF. These results suggest that scoparone can induce neurite outgrowth by stimulating the upstream steps of ERK, PKA, PKC and CaMK II in PC12 cells.
Sigma-Aldrich
6,7-Dimethoxycoumarin, 98%