MilliporeSigma
Use of admixtures in organic-contaminated cement-clay pastes.
Journal of hazardous materials (2008-06-03)
Paola Gallo Stampino, Luca Zampori, Giovanni Dotelli, Paola Meloni, Isabella Natali Sora, Renato Pelosato
ABSTRACT
In this work microstructure, porosity and hydration degree of cement-based solidified/stabilized wasteforms were studied before assessing their leaching behaviour. 2-Chloroaniline was chosen as a model liquid organic pollutant and included into cement pastes, which were also modified with different admixtures for concrete: a superplasticizer based on acrylic-modified polymer, a synthetic rubber latex and a waterproofing agent. An organoclay, modified with an ammonium quaternary salt (benzyl-dimethyl-tallowammonium, BDMTA), was added to the pastes as pre-sorbent agent of the organic matter. All the samples were dried up to constant weight in order to stop the hydration process at different times during the first 28 days of curing, typically, after 1 day (1d), 7 days (7d) and 28 days. Then, the microstructure of the hardened cement-clay pastes was investigated by powder X-ray diffraction (XRD). The hydration degree and porosity were studied by thermal analysis (TG/DTA) and mercury intrusion porosimetry (MIP), respectively. For samples cured for 28 days a short-term leach test set by Italian regulation for industrial waste recycling (D.M. 5 February 1998) was performed. The best results showed a 5% release of the total initial amount of organic pollutant.
Sigma-Aldrich
2-Chloroaniline, ≥99.5% (GC)
Sigma-Aldrich
2-Chloroaniline, technical, ≥98.0% (GC)