MilliporeSigma
  • Simultaneous spectrophotometric determination of mercury and palladium with Thio-Michler's Ketone using partial least squares regression and orthogonal signal correction.
Simultaneous spectrophotometric determination of mercury and palladium with Thio-Michler's Ketone using partial least squares regression and orthogonal signal correction.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-04-29)
Ali Niazi, Amir Azizi, Majid Ramezani
ABSTRACT
A simple, novel and sensitive spectrophotometric method was described for simultaneous determination of mercury and palladium. The method is based on the complex formation of mercury and palladium with Thio-Michler's Ketone (TMK) at pH 3.5. All factors affecting on the sensitivity were optimized and the linear dynamic range for determination of mercury and palladium found. The simultaneous determination of mercury and palladium mixtures by using spectrophotometric method is a difficult problem, due to spectral interferences. By multivariate calibration methods such as partial least squares (PLS), it is possible to obtain a model adjusted to the concentration values of the mixtures used in the calibration range. Orthogonal signal correction (OSC) is a preprocessing technique used for removing the information unrelated to the target variables based on constrained principal component analysis. OSC is a suitable preprocessing method for PLS calibration of mixtures without loss of prediction capacity using spectrophotometric method. In this study, the calibration model is based on absorption spectra in the 360-660 nm range for 25 different mixtures of mercury and palladium. Calibration matrices were containing 0.025-1.60 and 0.05-0.50 microg mL(-1) of mercury and palladium, respectively. The RMSEP for mercury and palladium with OSC and without OSC were 0.013, 0.006 and 0.048, 0.030, respectively. This procedure allows the simultaneous determination of mercury and palladium in synthetic and real matrix samples good reliability of the determination.
Sigma-Aldrich
4,4′-Bis(dimethylamino)thiobenzophenone, 85%