MilliporeSigma
  • A new method for N-N bond cleavage of N,N-disubstituted hydrazines to secondary amines and direct ortho amination of naphthol and its analogues.
A new method for N-N bond cleavage of N,N-disubstituted hydrazines to secondary amines and direct ortho amination of naphthol and its analogues.
Journal of the American Chemical Society (2008-04-15)
Qiang Tang, Chao Zhang, Meiming Luo
ABSTRACT
An unexpected reaction of N,N-disubstituted hydrazine with naphthol and its analogues under simply thermal conditions has been disclosed. 2-Naphthol reacted with various N,N-disubstituted hydrazines under argon to afford 1-amino-2-naphthol and the corresponding secondary amines in excellent yields. Ortho amination of 2-naphthols, hydroxyquinoline, and naphthalenamine occurred when they reacted with N-methyl-N-phenylhydrazine.
Sigma-Aldrich
7-Methoxy-2-naphthol, 98%
Sigma-Aldrich
p-Tolylhydrazine hydrochloride, 98%