MilliporeSigma
An expedient synthesis of mellitic triimides.
The Journal of organic chemistry (2008-04-12)
Kathryn G Rose, Deana M Jaber, Dina A Jaber, Chenaimwoyo A Gondo, Darren G Hamilton
ABSTRACT
Heating of the solid ammonium salts obtained from treatment of mellitic acid with 3 equiv of a primary amine yields trisubstituted mellitic triimides via dehydration and imide ring closure. This surprisingly simple synthetic approach is amenable to incorporation of alkyl, aryl, and amino acid ester substituents, thereby opening broad access to a family of C(3)-symmetric organic electron acceptors.
Sigma-Aldrich
Mellitic acid, 99%