MilliporeSigma
  • Enhancement of cutaneous nerve regeneration by 4-methylcatechol in resiniferatoxin-induced neuropathy.
Enhancement of cutaneous nerve regeneration by 4-methylcatechol in resiniferatoxin-induced neuropathy.
Journal of neuropathology and experimental neurology (2008-01-26)
Yu-Lin Hsieh, Hao Chiang, To-Jung Tseng, Sung-Tsang Hsieh
ABSTRACT
To generate an experimental neuropathy model in which small-diameter sensory nerves are specifically affected and to test a potential treatment, adult mice were given a single injection (50 microg/kg, i.p.) of the capsaicin analog resiniferatoxin (RTX). On Day 7 after RTX treatment, there was a 53% reduction in unmyelinated nerve density in the medial plantar nerve (p = 0.0067) and a 66% reduction in epidermal nerve density of hind paw skin (p = 0.0004) compared with vehicle-treated controls. Substance P-immunoreactive dorsal root ganglion neurons were also markedly depleted (p = 0.0001). These effects were associated with the functional deficit of prolonged withdrawal latencies to heat stimuli (p = 0.0007) on a hot plate test. The potential therapeutic effects of 4-methylcatechol (4MC) on this neuropathy were then tested by daily injections of 4MC (10 microg/kg, i.p.) from Days 7 to 35 after neuropathy induction. On Day 35, 4MC-treated mice had an increase in unmyelinated (p = 0.014) and epidermal nerve (p = 0.0013) densities and a reduction in thermal withdrawal latency (p = 0.0091) compared with RTX-only controls. These results indicate that 4MC promoted regeneration of unmyelinated nerves in experimental RTX-induced neuropathy and enhanced function.
Sigma-Aldrich
4-Methylcatechol, ≥95%