MilliporeSigma
  • Concise enantioselective total syntheses of (+)-homochelidonine, (+)-chelamidine, (+)-chelidonine, (+)-chelamine and (+)-norchelidonine by a Pd II-catalyzed ring-opening strategy.
Concise enantioselective total syntheses of (+)-homochelidonine, (+)-chelamidine, (+)-chelidonine, (+)-chelamine and (+)-norchelidonine by a Pd II-catalyzed ring-opening strategy.
Chemistry (Weinheim an der Bergstrasse, Germany) (2008-01-18)
Matthew J Fleming, Helen A McManus, Alena Rudolph, Walter H Chan, Jérémy Ruiz, Chris Dockendorff, Mark Lautens
ABSTRACT
New enantioselective syntheses of the B/C hexahydrobenzo[c]phenanthridine alkaloids (+)-homochelidonine, (+)-chelamidine, (+)-chelidonine, (+)-chelamine, and (+)-norchelidonine are described. Our rapid and convergent route to this class of natural products involved the development and application of a Pd II-catalyzed asymmetric ring-opening reaction of a meso-azabicyclic alkene with an aryl boronic acid as the key step. By screening a variety of functionalized ortho-substituted aryl boronic acids, chiral ligands and reaction conditions we were able to prepare the requisite cis-1-amino-2-aryldihydronaphthalenes in high yield and in up to 90 % ee. Early attempts to complete the synthesis of (+)-homochelidonine using an N-Boc azabicyclic alkene are described in full. The successful route required a protecting group alteration followed by B ring formation and then stereoselective installation of the C-11 syn-hydroxy group by regioselective epoxide ring-opening using a hydride source. Ring-opening of the same epoxide intermediate with water ultimately led to the synthesis of (+)-chelamidine. The same strategy was then used to synthesize the other structurally similar B/C hexahydrobenzo[c]phenanthridine alkaloids, (+)-chelidonine, (+)-chelamidine, and (+)-norchelidonine.
Sigma-Aldrich
Chelidonine, ≥97.0% (HPLC)