MilliporeSigma
  • Hepatoprotective potential of Fumaria indica Pugsley whole plant extracts, fractions and an isolated alkaloid protopine.
Hepatoprotective potential of Fumaria indica Pugsley whole plant extracts, fractions and an isolated alkaloid protopine.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2008-01-01)
Anshu Rathi, Arvind Kumar Srivastava, Annie Shirwaikar, Ajay Kumar Singh Rawat, Shanta Mehrotra
ABSTRACT
The present investigation demonstrates the hepatoprotective potential of 50% ethanolic water extract of whole plant of Fumaria indica and its three fractions viz., hexane, chloroform and butanol against d-galactosamine induced hepatotoxicity in rats. The hepatoprotection was assessed in terms reduction in histological damage, changes in serum enzymes (SGOT, SGPT, ALP) and metabolites bilirubin, reduced glutathione (GSH) and lipid peroxidation (MDA content). Among fractions more than 90% protection was found with butanol fraction in which alkaloid protopine was quantified as highest i.e. about 0.2mg/g by HPTLC. The isolated protopine in doses of 10-20mg p.o. also proved equally effective hepatoprotectants as standard drug silymarine (single dose 25mg p.o.). In general all treatments excluding hexane fraction proved hepatoprotective at par with silymarine (p<or=0.01).
Sigma-Aldrich
Protopine hydrochloride, ≥98%, solid