MilliporeSigma
  • Total synthesis of 7-deoxypancratistatin-1-carboxaldehyde and carboxylic acid via solvent-free intramolecular aziridine opening: phenanthrene to phenanthridone cyclization strategy.
Total synthesis of 7-deoxypancratistatin-1-carboxaldehyde and carboxylic acid via solvent-free intramolecular aziridine opening: phenanthrene to phenanthridone cyclization strategy.
Organic letters (2008-01-01)
Jonathan Collins, Melissa Drouin, Xuetong Sun, Uwe Rinner, Tomas Hudlicky
ABSTRACT
Solid-state silica-gel-catalyzed opening of aziridine 6 provided phenanthrene 7, whose oxidative cleavage, recyclization, and further elaboration furnished the C-1 aldehyde and carboxylic acid derivatives of 7-deoxypancratistatin for potential analogue synthesis.
Sigma-Aldrich
6-(5H)-Phenanthridinone, technical grade
Pricing and availability is not currently available.