MilliporeSigma
  • Development and validation of a method for the analysis of cafenstrole and its metabolite in brown rice grains and rice straw using high-performance liquid chromatography.
Development and validation of a method for the analysis of cafenstrole and its metabolite in brown rice grains and rice straw using high-performance liquid chromatography.
Biomedical chromatography : BMC (2007-12-07)
A M Abd El-Aty, Go-Woon Lee, M I R Mamun, Jeong-Heui Choi, Soon-Kil Cho, Ho-Chul Shin, Jae-Han Shim
ABSTRACT
The present work reports the extraction and clean-up procedures, as well as the chromatographic conditions developed, for the determination of cafenstrole and its metabolite (CHM-03) residues in brown rice grains and rice straw using HPLC-UV detection. The method makes use of an Apollo C(18) column and acetonitrile : water : acetic acid as a mobile phase for both cafenstrole and its metabolite in rice and rice straw. Using these conditions cafenstrole and its metabolite were resolved with a retention time (R(t)) of less than 14 min. The analytes were confirmed using positive atmospheric pressure ionization LC-MS with selected ion monitoring. The average recoveries of cafenstrole were found to be 87.0-92.5 and 87.6-88.3%. However, they ranged from 81.5 to 81.6% and from 76.1 to 78.5% for cafenstrole metabolite (CHM-03), in rice grains and rice straw, respectively, with relative standard deviations ranging from 1.4 to 6.6%. The limits of detection (LODs) of both cafenstrole and its metabolite were 0.002 and 0.02 ppm and 0.025 and 0.04 ppm, respectively. Field trials with recommended or double the recommended dose revealed that the herbicide could safely be recommended for application in rice and rice straw as no residues were detected in the harvest samples.