MilliporeSigma
  • [Determination of curcumol in plasma by HPLC-MS/MS method and its pharmacokinetics in Beagle dogs].
[Determination of curcumol in plasma by HPLC-MS/MS method and its pharmacokinetics in Beagle dogs].
Yao xue xue bao = Acta pharmaceutica Sinica (2007-12-07)
Rui Zhang, Ben-Jie Wang, Heng-Li Zhao, Xiao-Li Li, Chun-Min Wei, Rui-Chen Guo
ABSTRACT
To establish a high performance liquid chromatography (HPLC) coupled with tandem mass spectrometry quantitative detection method for the determination of curcumol, the main ingredient of zedoary turmeric oil fat emulsion, and investigate its pharmacokinetics in Beagle dogs, nine healthy Beagle dogs were divided into three groups, and blood samples were collected at scheduled time points after intravenous injection of 7.5, 10 and 12.5 mg x kg(-1) zedoary turmeric oil fat emulsion. The concentrations of curcumol were determined and pharmacokinetics was calculated. A good linearity was obtained from 0.25 to 100 ng x mL(-1) in plasma. The relative recoveries were from 91.33% to 103.17%, and the absolute recoveries were from 31.61% to 37.20%. The intra-day and inter-day variances (RSD) were < 15%. The main pharmacokinetic parameters of curcumol after intravenous injection of 7.5, 10 and 12.5 mg x kg(-1) zedoary turmeric oil fat emulsion were as follows, T1/2 : (2.0 +/- 0.4), (1.7 +/- 0.2) and (2.3 +/- 0.8) h, AUC(0-infinity): (15.1 +/- 2.7), (18.3 +/- 2.0) and (29.5 +/- 4.0) ng x mL(-1) x h; MRT: (0.9 +/- 0.1), (0.8 +/- 0.2) and (0.8 +/- 0.1) h, CL: (21.9 +/- 4.0), (24.9 +/- 6.0) and (18.4 +/- 1.2) L x h(-1) x kg; Vd : (65.4 +/- 26.5), (62.0 +/- 13.4) and (61.2 +/- 19.8) L x kg(-1), respectively. The developed method was rapid, highly sensitive and specific and could be used in curcumol pharmacokinetic studies in vivo. A three-compartment model was best fit to the plasma concentration--time curves obtained in Beagle dogs and the plasma AUC was increased proportionally with doses.