MilliporeSigma
  • Docetaxel and exisulind in previously treated non-small cell lung cancer (NSCLC) patients: a multicenter, phase II clinical trial.
Docetaxel and exisulind in previously treated non-small cell lung cancer (NSCLC) patients: a multicenter, phase II clinical trial.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer (2007-10-03)
Glen J Weiss, Everett E Vokes, Paul A Bunn, Lyn Magree, Jason Rusk, Don Albert, Karen Kelly
ABSTRACT
This multicenter, phase II clinical trial was conducted to evaluate the activity of the combination of docetaxel and exisulind in advanced non-small cell lung cancer (NSCLC) patients who failed a prior platinum-containing regimen. Patients with measurable disease and adequate organ function received exisulind (250 mg) given orally, twice daily, and docetaxel (36 mg/m) administered intravenously on days 1, 8, and 15 of a 4-week cycle for up to six cycles. In the absence of disease progression or intolerable side effects, patients continued taking 250 mg of exisulind orally, twice daily. Thirty-three patients (median age 60 years; range 34-77; median performance status 1) were enrolled. There were no objective responses documented. Sixteen patients [48%, 95% confidence interval (CI): 31%-66%] had stable disease after 8 weeks of treatment. Median progression-free survival (PFS) was 2.1 months (95% CI: 1.5-3.2 months); median overall survival time was 8.0 months (range 0.2-25.9 months). Toxicity was moderate, with dose adjustment for adverse event/toxicity required for docetaxel or exisulind in 13 (39.3%) patients. Grade 3/4 lymphopenia, neutropenia, and anemia occurred in 48.5%, 12.1%, and 9.1% of patients, respectively. Grade 3 or greater toxicity was seen in 12.1%, 6.1%, and 3% of patients for nausea/vomiting, dyspnea, and abdominal pain, respectively. Treatment with exisulind and weekly docetaxel was not active in NSCLC patients who failed a prior platinum-containing regimen. Further study of this combination does not seem warranted.
Sigma-Aldrich
Sulindac sulfone, ≥94% (HPLC), (solid or powder)