MilliporeSigma
  • [Effects of elevated atmospheric CO2 concentration on endogenous hormones in gingko leaves].
[Effects of elevated atmospheric CO2 concentration on endogenous hormones in gingko leaves].
Ying yong sheng tai xue bao = The journal of applied ecology (2007-09-25)
Xue-Mei Li, Xing-Yuan He, Wei Chen, Shi-Lei Fu, Li-Hong Zhang
ABSTRACT
With top-opened chamber system, this paper studied the responses of endogenous hormones abscisic acid (ABA), indoleacetic acid (IAA), zeatin riboside (ZR) and gibberellins (GA3) in gingko leaves to elevated atmospheric CO2 concentration (ambient + 350 micromol x mol(-1), EC). The results showed that EC could decrease the ABA content. After treated with EC for 120 days, the ABA content could be decreased by 63.0%, in comparing with the control. EC could also decrease the contents of IAA and ZR, and the decrement could reach significant with the extension of the treatment. The IAA content was 2 times higher after 100 days treatment, and ZR content was 2.5 times higher after 80 days treatment than those of CK. Under EC, the peak of GA3 appeared earlier. The (IAA + GA3 + ZR)/ABA ratio decreased gradually with the growth of gingko, and was obviously higher than the control at later stage (after 40-60 days treatment), indicating that elevated atmospheric CO2 concentration could promote the growth and development of gingko.
Sigma-Aldrich
trans-Zeatin-riboside, ~95%
Sigma-Aldrich
trans-Zeatin-riboside, suitable for plant cell culture, BioReagent, ~95%