MilliporeSigma
  • The DUF81 protein TauE in Cupriavidus necator H16, a sulfite exporter in the metabolism of C2 sulfonates.
The DUF81 protein TauE in Cupriavidus necator H16, a sulfite exporter in the metabolism of C2 sulfonates.
Microbiology (Reading, England) (2007-09-05)
Sonja Weinitschke, Karin Denger, Alasdair M Cook, Theo H M Smits
ABSTRACT
The degradation of taurine, isethionate and sulfoacetate in Cupriavidus necator (Ralstonia eutropha) H16 was shown by enzyme assays to be inducible, and each pathway involved sulfoacetaldehyde, which was subject to phosphatolysis by a common sulfoacetaldehyde acetyltransferase (Xsc, H16_B1870) to yield acetyl phosphate and sulfite. The neighbouring genes encoded phosphate acetyltransferase (Pta, H16_B1871) and a hypothetical protein [domain of unknown function (DUF)81, H16_B1872], with eight derived transmembrane helices. RT-PCR showed inducible transcription of these three genes, and led to the hypothesis that H16_B1872 and orthologous proteins represent a sulfite exporter, which was named TauE.