MilliporeSigma
  • Mouse liver effects of cyproconazole, a triazole fungicide: role of the constitutive androstane receptor.
Mouse liver effects of cyproconazole, a triazole fungicide: role of the constitutive androstane receptor.
Toxicological sciences : an official journal of the Society of Toxicology (2007-06-15)
Richard C Peffer, Jonathan G Moggs, Timothy Pastoor, Richard A Currie, Jayne Wright, Gill Milburn, Felix Waechter, Ivan Rusyn
ABSTRACT
Cyproconazole, a triazole fungicide, causes hepatocellular adenomas and carcinomas in CD-1 mice at dose levels of 100 and 200 ppm. The constitutive androstane receptor (CAR) has been shown to play a significant role in the overall mode of action for several nongenotoxic rodent carcinogens such as phenobarbital. The liver effects of dietary cyproconazole or phenobarbital were investigated after 2, 7, or 14 days in male CD-1, C57BL/6J, and C3H/HeNClrBR mice. Cyproconazole produced similar, dose-responsive effects in all three strains of mice, and the response was similar to that of phenobarbital. Subsequently, Car-null and wild-type male mice on a C3H/HeNClrBR background were administered 200 or 450 ppm cyproconazole, or 850 ppm phenobarbital for up to 7 days. In wild-type mice, 200 ppm cyproconazole caused liver hypertrophy, increased liver weight and cell proliferation, single-cell necrosis and fat vacuolation, effects generally similar to those caused by 850 ppm phenobarbital. Plasma cholesterol was decreased by both compounds, but cyproconazole had a greater effect. The higher dose (450 ppm) of cyproconazole caused similar changes, but greater evidence of liver damage was observed, including a large increase in plasma transaminases. Induction of CAR target genes Cyp2b10 and Gadd45beta was observed with both compounds, whereas the cell cycle regulatory gene Mdm2 was unaffected. In Car-null mice, the effects noted with either cyproconazole or phenobarbital were absent or greatly diminished. These experiments demonstrate that short-term liver effects of cyproconazole in mice are CAR-dependent and similar to those of phenobarbital, a known nongenotoxic rodent liver carcinogen.
Product Description
Supelco
Cyproconazol, PESTANAL®, analytical standard