MilliporeSigma
  • Conformational analysis of the electron-transfer kinetics across oligoproline peptides using N,N-dimethyl-1,4-benzenediamine donors and pyrene-1-sulfonyl acceptors.
Conformational analysis of the electron-transfer kinetics across oligoproline peptides using N,N-dimethyl-1,4-benzenediamine donors and pyrene-1-sulfonyl acceptors.
The journal of physical chemistry. B (2007-06-02)
Joseph B Issa, Abdu S Salameh, Edward W Castner, James F Wishart, Stephan S Isied
ABSTRACT
Photoinduced intramolecular charge separation across proline-bridged donor-acceptor complexes of the type Pyr-(Pro)n-DMPD (where Pyr=pyrene-1-sulfonyl and DMPD=N,N-dimethyl-1,4-phenylenediamine) was studied. The steady-state emission spectrum for n=0, 1, 2, 3 showed an increase in emission intensity with the number of proline residues. Time-dependent emission measured by streak camera showed increasing emission signal amplitude with increasing n, along with a decrease in decay rate. In all these studies, Pyr-Pro was used as a control complex for the decay of the excited pyrene acceptor moiety without the donor DMPD. Detailed photon counting experiments carried out in DMF/water, DMF, and toluene showed single-exponential kinetics for n=0, 1 and multiexponential kinetics for n=2, 3. Rate constants observed in DMF are for n=0, k=approximately 5x10(10) s(-1); n=1, k=9.70x10(8) s(-1); n=2, k=35.9x10(8) s(-1) (70%) and 5.58x10(8) s(-1) (30%); and n=3, k=16.6x10(8) s(-1) (55%) and 3.87x10(8) s(-1) (45%). These results show that a significant percentage of the n=2 and n=3 molecules undergo faster electron transfer than for the n=1 case. Conformational analysis for Pyr-(Pro)n-DMPD molecules in water showed that whereas only one conformation is possible for n=1, eight are possible for n=2, and 32 are possible for n=3. Calculation of the free energy and electronic coupling for these conformers in water showed that only a few of these conformations have the appropriate energy and electronic coupling to be observed in the experimental time window from 20 ps to 20 ns. Assignment of the conformers undergoing electron transfer in Pyr-(Pro)n-DMPD for n=2 and 3 was based on the values for the n=1 case, for which the measured rate constant is approximately 10(9) s(-1) and the calculated electronic coupling matrix element Hda is 297 cm(-1). The similarity in ground state energy between the cis and trans conformers for n=2 and 3, their use in aqueous-organic and organic solvents, and the nature of the Pyr and DMPD acceptor and donor groups could be contributing causes for the multiexponential kinetics, which was not observed for the metal ion derivatives of proline peptides studied earlier in aqueous solution.
Sigma-Aldrich
N,N-Dimethyl-p-phenylenediamine sulfate salt, 98%
Sigma-Aldrich
N,N-Dimethyl-p-phenylenediamine, 97%
Sigma-Aldrich
N,N-Dimethyl-1,4-phenylenediamine oxalate, 98%
Sigma-Aldrich
N,N-Dimethyl-p-phenylenediamine dihydrochloride, suitable for peroxidase test, ≥99.0% (titration)
Millipore
N,N-Dimethyl-p-phenylenediamine dihydrochloride, suitable for microbiology, ≥99.0%