MilliporeSigma
  • Visualization of hepatobiliary excretory function by intravital multiphoton microscopy.
Visualization of hepatobiliary excretory function by intravital multiphoton microscopy.
Journal of biomedical optics (2007-03-09)
Yuan Liu, Hsiao-Ching Chen, Shu-Mei Yang, Tzu-Lin Sun, Wen Lo, Ling-Ling Chiou, Guan Tarn Huang, Chen-Yuan Dong, Hsuan-Shu Lee
ABSTRACT
Intravital imaging of hepatobiliary excretion is vital for elucidating liver metabolism. In this work, we describe a novel method to observe the intravital dynamics of the uptake, processing, and excretion of an organic anion, 6-carboxyfluorescein diacetate (6-CFDA) in the hepatobiliary system. This is achieved by the use of multiphoton microscopy and an intravital hepatic imaging chamber. The high-quality images show sequential uptake and processing of 6-CFDA from the hepatocytes and the subsequent excretion into bile canaliculi within approximately 50 min. This is a promising technique to study intravital hepatic physiology and metabolism.
Sigma-Aldrich
6-Carboxyfluorescein diacetate, ≥95% (HPLC)