MilliporeSigma
  • Palladium-catalyzed benzene arylation: incorporation of catalytic pivalic acid as a proton shuttle and a key element in catalyst design.
Palladium-catalyzed benzene arylation: incorporation of catalytic pivalic acid as a proton shuttle and a key element in catalyst design.
Journal of the American Chemical Society (2006-12-21)
Marc Lafrance, Keith Fagnou
ABSTRACT
A palladium-pivalic acid cocatalyst system has been developed that exhibits unprecedented reactivity in direct arylation. This reactivity is illustrated with the first examples of high yielding direct metalation-arylation reactions of a completely unactivated arene, benzene. Experimental and computational evidence indicates that the pivalate anion is a key component in the palladation/C-H bond breaking event, that it lowers the energy of C-H bond cleavage and acts as a catalytic proton shuttle from benzene to the stoichiometric carbonate base. Eight examples of substituted aryl bromides are included which undergo direct arylation with benzene in 55-85% yield.
Sigma-Aldrich
Pivalic acid, 99%