MilliporeSigma
  • Comparison of three gas chromatography methods for the determination of slip agents in polyethylene films.
Comparison of three gas chromatography methods for the determination of slip agents in polyethylene films.
Journal of chromatography. A (2006-12-13)
Alvaro Garrido-López, Vanesa Esquiu, María Teresa Tena
ABSTRACT
Polymers require the use of some slip agents, such as oleamide and erucamide, in order to reduce their friction coefficient and to make films easier to handle. In this communication, three analytical methods consisting in pressurized liquid extraction (PLE) and gas chromatography (GC) are used to determine oleamide and erucamide in polyethylene films. The sample was extracted with pure isopropanol (two times) at 105 degrees C for 16 min. Then, the liquid extract containing oleamide and erucamide was analyzed by GC and three different detection systems: flame ionization detector (FID), thermoionic selective detector (TSD) and ion-trap mass spectrometry detector (MSD). Oleamide and erucamide were separated using a 30 m x 0.25 mm (I.D.) 5% phenyl-95% dimethyl-polysiloxane capillary column in 12 min. The chromatographic methods were characterized and compared in terms of repeatability, linearity and sensitivity. The GC-FID and GC-TSD methods were linear up to about 60 microg ml(-1), whereas the linear range for the GC-MSD method was shorter, from 20.5 to 42 microg ml(-1). LODs identified with GC-MSD were two times higher than those identified with the other two methods. Repeatability values (expressed as relative standard deviation) of less than 2.5% were found for FID and TSD but they were above 10% for MSD. Finally, each method was applied to determine the content of erucamide and oleamide in several polyethylene films and the results obtained were compared with those obtained from the nitrogen content measured by pyrolysis and gas-phase chemiluminescence. No significant differences were observed between the results of the methods.
Sigma-Aldrich
cis-13-Docosenoamide