MilliporeSigma
Oxidation studies on Andrographolide.
Natural product research (2006-11-28)
Vijayavitthal T Mathad, Satish Kumar, Kanwal Raj
ABSTRACT
Jones oxidation of Andrographolide (1), gave mixture of three products (3-dehydroandrographolide (5), 3,19-bis dehydroandrographolide (6) and 19-dehydroandrographolide (7). Tritylation of andrographolide at C19-OH resulted to products 8 and diene 9, which can be converted to its acetate 10 and oxidation product 11.
Sigma-Aldrich
Trityl chloride, purum, ≥97.0% (HPLC), ≥97.0% (AT)
Sigma-Aldrich
Trityl chloride, 97%