MilliporeSigma
  • 2-aminopyrimidine-4,6-diol as an efficient ligand for solvent-free copper-catalyzed N-arylations of imidazoles with aryl and heteroaryl halides.
2-aminopyrimidine-4,6-diol as an efficient ligand for solvent-free copper-catalyzed N-arylations of imidazoles with aryl and heteroaryl halides.
The Journal of organic chemistry (2006-10-10)
Ye-Xiang Xie, Shao-Feng Pi, Jian Wang, Du-Lin Yin, Jin-Heng Li
ABSTRACT
Efficient and solvent-free copper-catalyzed N-arylations of imidazoles with aryl and heteroaryl halides have been demonstrated. In the presence of CuBr, 2-aminopyrimidine-4,6-diol, and TBAF (n-Bu4NF), a variety of imidazoles underwent the N-arylation reaction with aryl and heteroaryl halides smoothly in moderate to excellent yields. Noteworthy is that the reaction is conducted under solvent-free conditions.
Sigma-Aldrich
2-Aminopyrimidine, 97%