MilliporeSigma
  • Role of the L-citrulline/L-arginine cycle in iNANC nerve-mediated nitric oxide production and airway smooth muscle relaxation in allergic asthma.
Role of the L-citrulline/L-arginine cycle in iNANC nerve-mediated nitric oxide production and airway smooth muscle relaxation in allergic asthma.
European journal of pharmacology (2006-08-22)
Harm Maarsingh, John Leusink, Johan Zaagsma, Herman Meurs
ABSTRACT
Nitric oxide synthase (NOS) converts L-arginine into nitric oxide (NO) and L-citrulline. In NO-producing cells, L-citrulline can be recycled to L-arginine in a two-step reaction involving argininosuccinate synthase (ASS) and -lyase (ASL). In guinea pig trachea, L-arginine is a limiting factor in neuronal nNOS-mediated airway smooth muscle relaxation upon inhibitory nonadrenergic noncholinergic (iNANC) nerve stimulation. Moreover, in a guinea pig model of asthma iNANC nerve-induced NO production and airway smooth muscle relaxation are impaired after the allergen-induced early asthmatic reaction, due to limitation of L-arginine. Using guinea pig tracheal preparations, we now investigated whether (i) the L-citrulline/L-arginine cycle is active in airway iNANC nerves and (ii) the NO deficiency after the early asthmatic reaction involves impaired L-citrulline recycling. Electrical field stimulation-induced relaxation was measured in tracheal open-rings precontracted with histamine. L-citrulline as well as the ASL inhibitor succinate did not affect electrical field stimulation-induced relaxation under basal conditions. However, reduced relaxation induced by a submaximal concentration of the NOS inhibitor N(omega)-nitro-L-arginine was restored by L-citrulline, which was prevented by the additional presence of succinate or the ASS inhibitor alpha-methyl-D,L-aspartate. Remarkably, the impaired iNANC relaxation after the early asthmatic reaction was restored by L-citrulline. In conclusion, the L-citrulline/L-arginine cycle is operative in guinea pig iNANC nerves in the airways and may be effective under conditions of low L-arginine utilization by nNOS (caused by NOS inhibitors), and during reduced L-arginine availability after allergen challenge. Enzymatic dysfunction in the L-citrulline/L-arginine cycle appears not to be involved in the L-arginine limitation and reduced iNANC activity after the early asthmatic reaction.