MilliporeSigma
  • Regiocontrolled synthesis of pyrrole-2-carboxaldehydes and 3-pyrrolin-2-ones from pyrrole Weinreb amides.
Regiocontrolled synthesis of pyrrole-2-carboxaldehydes and 3-pyrrolin-2-ones from pyrrole Weinreb amides.
The Journal of organic chemistry (2006-08-12)
Aaron R Coffin, Michael A Roussell, Elina Tserlin, Erin T Pelkey
ABSTRACT
A regiocontrolled synthesis of 3,4-disubstituted pyrrole-2-carboxaldehydes was completed in two steps from acyclic starting materials. A Barton-Zard pyrrole synthesis between N-methoxy-N-methyl-2-isocyanoacetamide and alpha-nitroalkenes or beta-nitroacetates provided N-methoxy-N-methyl pyrrole-2-carboxamides (pyrrole Weinreb amides), which were converted into the corresponding pyrrole-2-carboxaldehydes by treatment with lithium aluminum hydride. A regioselective oxidation of the pyrrole-2-carboxaldehydes gave the corresponding 3,4-disubstituted 3-pyrrolin-2-ones.
Sigma-Aldrich
Pyrrole-2-carboxaldehyde, 98%